MilliporeSigma
  • New applications of sporopollenin as a solid phase support for peptide synthesis and the use of sonic agitation.
New applications of sporopollenin as a solid phase support for peptide synthesis and the use of sonic agitation.
International journal of peptide and protein research (1983-11-01)
R Adamson, S Gregson, G Shaw
ABSTRACT
The synthesis of a tetrapeptide on a base functionalised sporopollenin using sonic agitation for coupling and washing processes is described.