MilliporeSigma
  • Influence of membrane organization on the interactions between persistent pollutants and model membranes.
Influence of membrane organization on the interactions between persistent pollutants and model membranes.
Bioelectrochemistry (Amsterdam, Netherlands) (2012-01-03)
Dorota Matyszewska, Ewa Wypijewska, Renata Bilewicz
ABSTRACT
Langmuir monolayer studies and electrochemical methods were employed to investigate the effect of model membrane organization on the interactions with persistent pollutants such as perfluorinated carboxylic acids (PFCAs). 1,2-dimyristoyl-sn-glycero-3-phosphoethanoloamine (DMPE) was employed to construct the model lipid membrane and perfluorooctanesulphonic acid (PFOS) was chosen as the representative of perfluorinated pollutants. We demonstrate that perfluorinated compounds penetrate a model membrane only when it is less condensed. Such liquid-expanded phase was achieved by compressing the Langmuir monolayer to lower surface pressures. PFOS incorporation into model DMPE membrane during membrane formation was observed in liquid-expanded region, while at higher surface pressures, in the well-organized monolayer the expulsion of perfluorinated compound occurred as the result of a strong attraction between the DMPE molecules. The DMPE monolayers prepared by the Langmuir-Blodgett/Langmuir-Schaefer method were transferred onto gold electrode under surface pressure of 3 mN/m and 20 mN/m. The model membrane organization depends on surface pressure during transfer and time of exposure to PFOS solution and is shown to affect the electrode accessibility by three electroactive compounds used as the probes of the blocking properties of the monolayer: menadione, potassium ferricyanide and hexaamineruthenium chloride, differing in the properties and kinetics of electron transfer.
Sigma-Aldrich
Potassium ferricyanide(III), powder or chunks, <10 μm, 99%
Sigma-Aldrich
Potassium hexacyanoferrate(III), BioUltra, ≥99.0% (RT)
Sigma-Aldrich
Potassium hexacyanoferrate(III), 99.98% trace metals basis
Sigma-Aldrich
Potassium hexacyanoferrate(III), ReagentPlus®, ~99%
Sigma-Aldrich
Potassium hexacyanoferrate(III), ACS reagent, ≥99.0%