MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Glutamate dehydrogenase, insulin secretion, and type 2 diabetes: a new means to protect the pancreatic β-cell?
Glutamate dehydrogenase, insulin secretion, and type 2 diabetes: a new means to protect the pancreatic β-cell?
The Journal of endocrinology (2012-01-11)
Isabel Göhring, Hindrik Mulder
ABSTRACT
In this issue of Journal of Endocrinology, Dr Han and colleagues report a protective effect of the glutamate dehydrogenase activator 2-aminobicyclo-(2,2,1)-heptane-2-carboxylic acid (BCH) under diabetes-like conditions that impair β-cell function in both a pancreatic β-cell line and db/db mice. Based on these observations, the authors suggest that BCH could serve as a novel treatment modality in type 2 diabetes. The present commentary discusses the importance of the findings. Some additional questions are raised, which may be addressed in future investigations, as there is some concern regarding the BCH treatment of β-cell failure.
Sigma-Aldrich
2-Amino-2-norbornanecarboxylic acid, amino acid transport inhibitor