MilliporeSigma
  • Pharmacokinetic and bioequivalence study of etoricoxib tablet in healthy Bangladeshi volunteers.
Pharmacokinetic and bioequivalence study of etoricoxib tablet in healthy Bangladeshi volunteers.
Arzneimittel-Forschung (2012-01-12)
M Hasanuzzaman Shohag, Mohammad Safiqul Islam, Maizbha Uddin Ahmed, Jafreen Jamal Joti, M Siddiqul Islam, M Hasanuzzaman, Abul Hasnat
ABSTRACT
The aim of this study was to compare the pharmacokinetic properties of two etoricoxib (CAS 202409-33-4) 60 mg formulations, namely Etocox-60 (test product) and reference product, and to evaluate whether these two formulations meet the FDA criteria to assume bioequivalence. Twenty-four healthy volunteers were enrolled into this randomized, single-dose, 2-way crossover, open-label pharmacokinetic study. Subjects were randomly assigned to receive the test formulation followed by the reference formulation or vice versa as a single dose of 60 mg tablets after 12 h overnight fasting, with a washout period of two weeks. Following oral administration, blood samples were collected at 0 (baseline), 0.5, 1.0, 1.5, 2.0, 3.0, 4.0, 6.0, 8.0, 10.0, 12.0, 24.0, 48.0, 72.0, 96.0, and 120.0 h. Serum concentration of etoricoxib was assessed using a high performance liquid chromatographic-UV spectrometry procedure. The pharmacokinetic parameters were determined by the non-compartmental method. After administering a single dose of 60 mg of each etoricoxib formulation, the obtained mean (SD) values for the test and reference products were 1.26 (0.33) and 1.29 (0.35) microg/ml for Cmax; 3.25 (2.64) and 2.63 (1.40) h for t(max); 29.63 (8.31) and 30.40 (5.85) h x microg/ml for AUC0-120; and 31.84 (10.97) and 33.00 (8.10) h x microg/ml for AUC0-infinity, respectively. The mean t1/2 was found 27.99 (7.87) h and 29.84 (7.93) h for test and reference product respectively. From paired t-test, no significant differences were observed (p > 0.05) for any pharmacokinetic parameters. After analysis of variance, no period, sequence or formulation effects were observed for any pharmacokinetic property. The 90% confidence intervals of the test/reference mean ratios of the 1n-transformed AUC0-120, AUC0-infinity and Cmax mean values were 95.90% (85.37%-107.74 %), 94.69% (84.43%-106.20%) and 97.87% (85.54 %-111.98 %), respectively, which fell within the predetermined FDA bioequivalence range of 80%-125%. This single-dose study found that the test and reference formulations of etoricoxib met the regulatory criteria for bioequivalence in terms of both rate and extent of absorption.
Product Description
Supelco
Etoricoxib, VETRANAL®, analytical standard