MilliporeSigma
  • Rhodium-catalyzed 1,3-acyloxy migration and subsequent intramolecular [4+2] cycloaddition of vinylallene and unactivated alkyne.
Rhodium-catalyzed 1,3-acyloxy migration and subsequent intramolecular [4+2] cycloaddition of vinylallene and unactivated alkyne.
Chemical communications (Cambridge, England) (2012-01-19)
Suyu Huang, Xiaoxun Li, Claire L Lin, Ilia A Guzei, Weiping Tang
ABSTRACT
A Rh-catalyzed 1,3-acyloxy migration of propargyl ester followed by intramolecular [4+2] cycloaddition of vinylallene and unactivated alkyne was developed. This tandem reaction provides access to bicyclic compounds containing a highly functionalized isotoluene or cyclohexenone structural motif, while only aromatic compounds were observed in related transition metal-catalyzed cycloadditions.
Sigma-Aldrich
Allene, ≥95%