MilliporeSigma
  • Tinea of the scrotum--report of a case presenting as lichenified plaques.
Tinea of the scrotum--report of a case presenting as lichenified plaques.
The Journal of dermatology (1990-07-01)
S Dekio, Q Li-mo, Y Kawasaki, J Jidoi
ABSTRACT
A 57-year-old Japanese man with tinea of the scrotum was described. His lesions on the scrotum were unusual; they appeared as lichenified plaques.