MilliporeSigma
  • Odor abatement in biotrickling filters: effect of the EBRT on methyl mercaptan and hydrophobic VOCs removal.
Odor abatement in biotrickling filters: effect of the EBRT on methyl mercaptan and hydrophobic VOCs removal.
Bioresource technology (2012-02-09)
Raquel Lebrero, Elisa Rodríguez, José M Estrada, Pedro A García-Encina, Raúl Muñoz
ABSTRACT
The performance and microbiology of a biotrickling filter (BTF) treating methyl mercaptan, toluene, alpha-pinene and hexane at the mg m(-3) level was studied at empty bed residence times (EBRT) of 50, 30, 11 and 7 s. Removal efficiencies (REs) higher than 95% were observed for MeSH, toluene and alpha-pinene even at 11 s, while hexane REs exceeded 70%. At 7 s, an irreversible damage of the microbial activity due to the accumulation of toxic metabolites resulted in a decrease of REs. The addition of silicone stabilized process performance but only re-inoculation allowed achieving a complete removal of MeSH, toluene and alpha-pinene, and hexane REs of 80%. The high K(L)a values (ranging from 38 ± 4 to 90 ± 11 h(-1)) explained the good BTF performance at such low EBRTs. A high bacterial diversity, along with a vertical distribution of the bacterial communities was observed, the main phyla being Proteobacteria, Actinobacteria, Nitrospira, Chloroflexi and Gemmatimonadertes.
Sigma-Aldrich
Sodium thiomethoxide, 95%
Sigma-Aldrich
Sodium methanethiolate, technical, ≥90% (RT)