MilliporeSigma
  • Negative chemical ionization GC/MS determination of nitrite and nitrate in seawater using exact matching double spike isotope dilution and derivatization with triethyloxonium tetrafluoroborate.
Negative chemical ionization GC/MS determination of nitrite and nitrate in seawater using exact matching double spike isotope dilution and derivatization with triethyloxonium tetrafluoroborate.
Analytical chemistry (2012-02-11)
Enea Pagliano, Juris Meija, Ralph E Sturgeon, Zoltan Mester, Alessandro D'Ulivo
ABSTRACT
The alkylation of nitrite and nitrate by triethyloxonium tetrafluoroborate allows determination of their ethyl esters by headspace gas chromatography/mass spectrometry (GC/MS). In the present study, significant improvement in analytical performance is achieved using negative chemical ionization providing detection limits of 150 ng/L for NO(2)(-) and 600 ng/L for NO(3)(-), an order of magnitude better than those achieved using electron impact ionization. The derivatization procedure was optimized and alkaline conditions adopted to minimize conversion of nitrite to nitrate (determined to be 0.07% at 100 mg/L NO(2)(-)) and to avoid the exchange of oxygen between the analytes and the solvent (water). Quantitation entails use of isotopically enriched standards (N(18)O(2)(-) and (15)NO(3)(-)), which also permits monitoring of potential conversion from nitrite to nitrate during the analysis (double spike isotope dilution).
Sigma-Aldrich
Triethyloxonium tetrafluoroborate, ≥97.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Triethyloxonium tetrafluoroborate solution, 1.0 M in methylene chloride
Pricing and availability is not currently available.