MilliporeSigma
  • Highly sensitive fluorescent sensing for water based on poly(m-aminobenzoic acid).
Highly sensitive fluorescent sensing for water based on poly(m-aminobenzoic acid).
Chemical communications (Cambridge, England) (2012-02-14)
Qiliang Deng, Yanli Li, Jianhua Wu, Yang Liu, Guozhen Fang, Shuo Wang, Yukui Zhang
ABSTRACT
Fluorescent conjugated polymer, poly(3-aminobenzoic acid), as a new class of fluorescence sensor for detection of trace amounts of water in organic solvents was developed. This sensor exhibits extraordinarily high sensitivity with a detection limit as low as 0.008 wt% for water in DMF.
Sigma-Aldrich
3-Aminobenzoic acid, 98%