MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
High yielding synthesis of N-ethyl dehydroamino acids.
Amino acids (2012-02-22)
Luís S Monteiro, Ana S Suárez
ABSTRACT
Recently we reported the use of a sequence of alkylation and dehydration methodologies to obtain N-ethyl-α, β-dehydroamino acid derivatives. The application of this N-alkylation procedure to several methyl esters of β,β-dibromo and β-bromo, β-substituted dehydroamino acids protected with standard amine protecting groups was subsequently reported. The corresponding N-ethyl, β-bromo dehydroamino acid derivatives were obtained in fair to high yields and some were used as substrates in Suzuki cross-coupling reactions to give N-ethyl, β,β-disubstituted dehydroalanine derivatives. Herein, we further explore N-ethylation of β-halo dehydroamino acid derivatives using triethyloxonium tetrafluoroborate as alkylating agent, but substituting N,N-diisopropylethylamine for potassium tert-butoxide as auxiliary base. In these conditions, for all β-halo dehydroamino acid derivatives, reactions were complete and the N-ethylated derivative could be isolated in high yield. This method was also applied for N-ethylation of non-halogenated dehydroamino acids. Again, with all compounds the reactions were complete and the N-ethyl dehydroamino acid derivatives could be isolated in high yields. Some of these N-ethyl dehydroamino acid methyl ester derivatives were converted in high yields to their corresponding acids and coupled to an amino acid methyl ester to give N-ethyl dehydrodipeptide derivatives in good yields. Thus, this method constitutes a general procedure for high yielding synthesis of N-ethylated dehydroamino acids, which can be further applied in peptide synthesis.
Sigma-Aldrich
Triethyloxonium tetrafluoroborate, ≥97.0% (T)
Sigma-Aldrich
Triethyloxonium tetrafluoroborate solution, 1.0 M in methylene chloride
Sigma-Aldrich
Potassium tert-butoxide solution, 1.0 M in THF
Sigma-Aldrich
Potassium tert-butoxide, reagent grade, ≥98%
Sigma-Aldrich
Potassium tert-butoxide, sublimed grade, 99.99% trace metals basis