MilliporeSigma
  • Growth and characterization of pure and glycine doped cadmium thiourea sulphate (GCTS) crystals.
Growth and characterization of pure and glycine doped cadmium thiourea sulphate (GCTS) crystals.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-02-22)
M Lawrence, J Thomas Joseph Prakash
ABSTRACT
The pure and glycine doped cadmium thiourea sulphate (GCTS) single crystals were grown successfully by slow evaporation method at room temperature. The concentration of dopant in the mother solution was 1 mol%. There is a change in unit cell. The Fourier transform infrared spectroscopy study confirms the incorporation of glycine into CTS crystal. The doped crystals are optically better and more transparent than the pure ones. The dopant increases the hardness value of the material. The grown crystals were also subjected to thermal and NLO studies.
Sigma-Aldrich
Cadmium sulfate 8/3-hydrate, puriss. p.a., ACS reagent, ≥99.0% (calc. based on CdSO4 · 8/3 H2O, KT)
Sigma-Aldrich
Cadmium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Cadmium sulfate 8/3-hydrate, SAJ super special grade, ≥99.0%