MilliporeSigma
  • Ion size influence on the Ar solvation shells of M(+)-C6F6 clusters (M = Na, K, Rb, Cs).
Ion size influence on the Ar solvation shells of M(+)-C6F6 clusters (M = Na, K, Rb, Cs).
The journal of physical chemistry. A (2012-03-02)
M Albertí, N Faginas Lago
ABSTRACT
The size-specific influence of alkali metal ions in the gradual transition from cluster rearrangement to solvation dynamics is investigated by means of molecular dynamics simulations for alkali metal cation-hexafluorobenzene systems, M(+)-C(6)F(6) (M = Na, K, Rb and Cs), surrounded by Ar atoms. To analyze such transition, different small aggregates of the M(+)-C(6)F(6)-Ar(n) (n = 1, ..., 30) type and M(+)-C(6)F(6) clusters solvated by about 500 Ar atoms are considered. The Ar-C(6)F(6) interaction contribution has been described using two different formalisms, based on the interaction decomposition in atom-bond and in atom-effective atom terms, which have been applied to study the small aggregates and to investigate the Ar solvated M(+)-C(6)F(6) clusters, respectively. The selectivity of the promoted phenomena from the M(+) ion size and their dependence from the number of Ar atoms is characterized.
Sigma-Aldrich
Hexafluorobenzene, 99%
Sigma-Aldrich
Hexafluorobenzene, ≥99.5%, NMR grade