MilliporeSigma
  • Sweet block copolymer nanoparticles: preparation and self-assembly of fully oligosaccharide-based amphiphile.
Sweet block copolymer nanoparticles: preparation and self-assembly of fully oligosaccharide-based amphiphile.
Biomacromolecules (2012-03-09)
Samuel de Medeiros Modolon, Issei Otsuka, Sébastien Fort, Edson Minatti, Redouane Borsali, Sami Halila
ABSTRACT
The preparation of biocompatible nanocarriers that have potential applications in the cosmetic and health industries is highly desired. The self-assembly of amphiphilic block copolymers displaying biosourced polysaccharides at the surface is one of the most promising approaches. In the continuity of our works related to the preparation of "hybrid" amphiphilic oligosaccharide-based block copolymers, we present here the design of a new generation of self-assembled nanoparticles composed entirely of oligosaccharide-based amphiphilic block co-oligomers (BCO). These systems are defined by a covalent linkage of the two saccharidic blocks through their reducing end units, resulting in a sweet "head-to-head" connection. As an example, we have prepared and studied a BCO in which the hydrophilic part is composed of a free maltoheptaosyl derivative clicked to a hydrophobic part composed of a peracetylated maltoheptaosyl derivative. This amphiphilic BCO self-assembles to form spherical micelles in water with an average diameter of 30 nm. The efficient enzymatic hydrolysis of the maltoheptaose that constitutes the shell of the micelles was followed by light scattering and colorimetric methods.
Sigma-Aldrich
Maltoheptaose, ≥60% (HPLC)