MilliporeSigma
  • Visible light-harvesting perylenebisimide-fullerene (C60) dyads with bidirectional "ping-pong" energy transfer as triplet photosensitizers for photooxidation of 1,5-dihydroxynaphthalene.
Visible light-harvesting perylenebisimide-fullerene (C60) dyads with bidirectional "ping-pong" energy transfer as triplet photosensitizers for photooxidation of 1,5-dihydroxynaphthalene.
Chemical communications (Cambridge, England) (2012-03-09)
Yifan Liu, Jianzhang Zhao
ABSTRACT
Visible light-harvesting perylenebisimide (PBI)-C(60) dyads were prepared as organic triplet photosensitizers for photooxidation of 1,5-dihydroxynaphthalene and the efficiency of the dyads is 6-fold of the conventional Ir(III) complex triplet photosensitizer.
Sigma-Aldrich
1,5-Dihydroxynaphthalene, 97%