MilliporeSigma
  • Comparative evaluation of the efficacy of three commercially available toothpastes on dentin hypersensitivity reduction: an eight-week clinical study.
Comparative evaluation of the efficacy of three commercially available toothpastes on dentin hypersensitivity reduction: an eight-week clinical study.
The Journal of clinical dentistry (2011-01-01)
Raffaella Docimo, Cesare Perugia, Martina Bartolino, Paolo Maturo, Luigi Montesani, Yun Po Zhang, William DeVizio, Luis R Mateo, Serge Dibart
ABSTRACT
The objective of this eight-week, single-center, three-cell, double-blind, and randomized clinical study was to evaluate the dentin hypersensitivity reduction efficacy of three commercially available toothpastes: 1) Colgate Sensitive Pro-Relief Toothpaste (also marketed as elmex Sensitive Professional); 2) Sensodyne Rapid Relief Toothpaste; and (3) Crest Cavity Protection Toothpaste. 150 subjects, having two teeth with tactile and air blast hypersensitivity, were assigned to one of the three study groups (50/group). Subjects were then asked to brush their teeth for one minute, twice daily, with the given toothpaste. The dentin hypersensitivity and oral tissues were evaluated at baseline, two weeks, four weeks, and eight weeks. Comparison of the treatment groups with respect to gender was conducted using a chi-square analysis, and with respect to age and baseline hypersensitivity scores was performed using the analysis of variance (ANOVA). Within-treatment effects were analyzed using the paired t-test, while the analysis of covariance (ANCOVA) was used to examine between-treatment effects. The post hoc Tukey test was performed for pair-wise comparisons. All statistical tests were two-sided using a significance level of alpha = 0.05. After two, four, and eight weeks of daily use of the products, all three groups showed a statistically significant reduction from baseline in tactile and air blast dentin hypersensitivity (p < 0.05). Colgate Sensitive Pro-Relief toothpaste produced a significant improvement in mean tactile and air blast dentin hypersensitivity scores, and was more effective than Sensodyne Rapid Relief toothpaste and Crest Cavity Protection toothpastes (p < 0.05). Colgate Sensitive Pro-ReliefToothpaste, used twice daily, significantly reduces dentin hypersensitivity, and is significantly more effective in reducing dentin hypersensitivity than Sensodyne Rapid Relief Toothpaste and Crest Cavity Protection Toothpaste.
Sigma-Aldrich
Strontium acetate, 99.95% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Strontium acetate
Pricing and availability is not currently available.