MilliporeSigma
Photochromism of a diarylethene having an azulene ring.
The Journal of organic chemistry (2012-03-14)
Jun-ichiro Kitai, Takao Kobayashi, Waka Uchida, Makoto Hatakeyama, Satoshi Yokojima, Shinichiro Nakamura, Kingo Uchida
ABSTRACT
Diarylethene derivatives incorporating an azulene ring at the ethene moiety were synthesized. One derivative having thiazole rings showed the expected coloration reaction by excitation at 313 nm (to a higher singlet state) but not when excited at 635 nm (S(0) to S(1) excitation). The system demonstrates that the cyclization reaction can be controlled by selective excitation at different wavelengths of the absorption spectrum. On the other hand, another derivative having thiophene rings did not show any photochromism. The results clearly show the importance of the coplanarity of the system for the photoisomerization.
Sigma-Aldrich
Azulene, 99%