MilliporeSigma
  • Solid-vapor sorption of xylenes: prioritized selectivity as a means of separating all three isomers using a single substrate.
Solid-vapor sorption of xylenes: prioritized selectivity as a means of separating all three isomers using a single substrate.
Angewandte Chemie (International ed. in English) (2012-03-14)
Matteo Lusi, Leonard J Barbour
ABSTRACT
A Werner complex is highly selective for o-xylene in a vapor mixture containing all three isomers. However, in the absence of o-xylene, the substrate shows similar selectivity for m-xylene over p-xylene. Kinetic studies show a different trend whereby m-xylene is absorbed most rapidly, implying that thermodynamic factors must be responsible for the selectivity.
Product Description
Supelco
m-Xylene, analytical standard
Supelco
m-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
m-Xylene, anhydrous, ≥99%
Sigma-Aldrich
m-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
m-Xylene, ReagentPlus®, 99%
Sigma-Aldrich
Xylenes, reagent grade
Sigma-Aldrich
Xylenes, histological grade
Sigma-Aldrich
Xylenes, ACS reagent, ≥98.5% xylenes + ethylbenzene basis
Supelco
p-Xylene, analytical standard
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
p-Xylene, anhydrous, ≥99%
Sigma-Aldrich
Xylenes, SAJ first grade, ≥80.0%
Sigma-Aldrich
p-Xylene, SAJ first grade, ≥99.0%
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Sigma-Aldrich
Xylenes, SAJ special grade, ≥80.0%
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
p-Xylene, ReagentPlus®, 99%
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
p-Xylene, suitable for HPLC, ≥99%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Supelco
p-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material