MilliporeSigma
  • Design and evaluation of poly(DL-lactic-co-glycolic acid) nanocomposite particles containing salmon calcitonin for inhalation.
Design and evaluation of poly(DL-lactic-co-glycolic acid) nanocomposite particles containing salmon calcitonin for inhalation.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2012-03-15)
Mingshi Yang, Hiromitsu Yamamoto, Homare Kurashima, Hirofumi Takeuchi, Toyokazu Yokoyama, Hiroyuki Tsujimoto, Yoshiaki Kawashima
ABSTRACT
Salmon calcitonin, for the treatment of calcium homeostasis and bone remodeling, was used as a model peptide drug and adsorbed on the surface of biodegradable polymeric poly(dl-lactic-co-glycolic acid) (PLGA) nanospheres. Subsequently, the nanospheres were treated using lyophilizer and loaded onto inhalable carrier using Mechanofusion to obtain nanocomposite particles suitable for inhalation. The physicochemical properties and in vitro inhalation properties of the nanocomposite particles were investigated. The pulmonary distribution and pharmacological effect were also evaluated in male Wistar rats. The results showed that the drug loading efficiency of salmon calcitonin on PLGA nanospheres were exceeding 96% (w/w). Inhalation efficiency of the lyophilized PLGA nanospheres was largely improved after they were loaded on the surface of inhalable carrier. Over 50% (w/w) of the lyophilized PLGA nanospheres could be deposited in the alveoli section after intratracheal administration to male Wistar rats, while a rapid elimination rate of the lyophilized nanospheres from the lung was found in pulmonary distribution study. The in vivo pharmacological study showed that the nanocomposite particles exhibited superior hypocalcemic action over salmon calcitonion solution and the lyophilized nanospheres. It suggested that the Mechanofusion(TM) technique can impart improved inhalation properties to the lyophilized nanospheres for pulmonary delivery of therapeutic peptide drugs.
Sigma-Aldrich
Calcitonin salmon, ≥97% (HPLC), powder
Calcitonin salmon, European Pharmacopoeia (EP) Reference Standard