MilliporeSigma
  • Regioselective Cu(I)-catalyzed tandem A3-coupling/decarboxylative coupling to 3-amino-1,4-enynes.
Regioselective Cu(I)-catalyzed tandem A3-coupling/decarboxylative coupling to 3-amino-1,4-enynes.
Organic letters (2012-03-30)
Huangdi Feng, Denis S Ermolat'ev, Gonghua Song, Erik V Van der Eycken
ABSTRACT
An efficient and novel copper-mediated protocol for the synthesis of 3-amino-1,4-enynes from glyoxylic acid, an amine, and an alkyne was developed. This new reaction involving two sequential C-C bond formations is air and moisture tolerant and proceeds via a tandem A(3)-coupling and a selective decarboxylative coupling.
Sigma-Aldrich
Glyoxylic acid solution, 50 wt. % in H2O