MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Forces between hydrophobic solids in concentrated aqueous salt solution.
Forces between hydrophobic solids in concentrated aqueous salt solution.
Physical review letters (2012-04-03)
Dean J Mastropietro, William A Ducker
ABSTRACT
Much research has focused on the discovery and description of long-ranged forces between hydrophobic solids immersed in water. Here we show that the force between high contact-angle solids in concentrated salt solution (1 M KCl) agrees very well with van der Waals forces calculated from Lifshitz theory for separations greater than 5 nm. The hydrophobic solids are octadecyltrichlorosilane-coated glass, with an advancing contact angle of 108°. Thus, in 1 M salt solution, it is unnecessary to invoke the presence of a hydrophobic force at separations greater than 5 nm. Through measurement in salt solution, we avoid the necessity of accounting for large electrostatic forces that frequently occur in pure water and may obscure resolution of other forces.
Sigma-Aldrich
Trichloro(octadecyl)silane, ≥90%