MilliporeSigma
  • A novel "pro-sensitizer" based sensing of enzymes using Tb(III) luminescence in a hydrogel matrix.
A novel "pro-sensitizer" based sensing of enzymes using Tb(III) luminescence in a hydrogel matrix.
Chemical communications (Cambridge, England) (2012-04-03)
Sandip Bhowmik, Uday Maitra
ABSTRACT
Chemically synthesized "pro-sensitizers" release the sensitizer in the presence of lipase or β-glucosidase, triggering a significant luminescence response from a lanthanide based hydrogel.
Sigma-Aldrich
2,3-Dihydroxynaphthalene, ≥98.0% (HPLC)