MilliporeSigma
  • Liver tumor promoting effect of etofenprox in rats and its possible mechanism of action.
Liver tumor promoting effect of etofenprox in rats and its possible mechanism of action.
The Journal of toxicological sciences (2012-04-03)
Yuri Hojo, Ayako Shiraki, Takuma Tsuchiya, Keisuke Shimamoto, Yuji Ishii, Kazuhiko Suzuki, Makoto Shibutani, Kunitoshi Mitsumori
ABSTRACT
To investigate the liver tumor-promoting effects of etofenprox (ETF), a pyrethroid-like insecticide, 6 week-old male F344 rats were given an intraperitoneal injection of N-diethylnitrosamine (DEN). After 2 weeks from the DEN treatment, 12 rats per group received a powdered diet containing 0, 0.25, 0.50, or 1.0% ETF for 8 weeks. At the time of 2nd week of ETF administration, all animals were subjected to two-thirds partial hepatectomy (PH). One rat per group except for the 0.25% ETF group died due to surgical operation of PH. The number and area of glutathione S-transferase placental form (GST-P) positive foci significantly increased in the livers of DEN-initiated rats given 0.50% and 1.0% ETF compared with the DEN-alone group. Quantitative real-time RT-PCR analysis revealed that the mRNA expression of phase I enzymes Cyp2b1/2, phase II enzymes such as Akr7a3, Gsta5, Ugt1a6, Nqo1 significantly increased in the DEN+ETF groups. The immunohistochemistry showed the translocation of CAR from the cytoplasm to the nuclei of hepatocytes in the ETF-treated groups. Reactive oxygen species (ROS) production increased in microsomes isolated from the livers of ETF-treated rats, and thiobarbituric acid-reactive substances (TBARS) levels and 8- hydroxy-2-deoxyguanosine (8-OHdG) content significantly increased in all of the ETF-treated groups and DEN+1.0% ETF group, respectively. The results of the present study indicate that ETF has a liver tumor-promoting activity in rats, and suggest that ETF activates the constitutive active/androstane receptor (CAR) and enhances microsomal ROS production, resulting in the upregulation of Nrf2 gene batteries; such an oxidative stress subsequently induces liver tumor-promoting effects by increased cellular proliferation.
Product Description
Supelco
Etofenprox, PESTANAL®, analytical standard