MilliporeSigma
Integrating techniques to restore an adolescent's smile.
Dentistry today (2012-04-10)
Ana Paula, Paulo Henrique dos Santos, Fernanda Garcia de Oliveira, Lucas Silveira Machado, Daniel Sundfeld Neto, Renato Herman Sundfeld