MilliporeSigma
  • Palladium-catalyzed asymmetric 6-endo cyclization of dienamides with substituent-driven activation.
Palladium-catalyzed asymmetric 6-endo cyclization of dienamides with substituent-driven activation.
Organic letters (2012-04-26)
Hiroshi Tsuchikawa, Yuya Maekawa, Shigeo Katsumura
ABSTRACT
Chiral 2-piperidinone compounds with various C-6 substituents were successfully synthesized via a Pd-catalyzed asymmetric 6-endo cyclization of dienamides, which were evidently activated by both N-p-toluenesulfonyl and C-3 ester substituents.
Sigma-Aldrich
δ-Valerolactam, 98%