MilliporeSigma
  • Large increase of the lifetime of a charge-separated state in a molecular triad induced by hydrogen-bonding solvent.
Large increase of the lifetime of a charge-separated state in a molecular triad induced by hydrogen-bonding solvent.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-04-26)
Jihane Hankache, Oliver S Wenger
Sigma-Aldrich
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
Supelco
1,1,1,3,3,3-Hexafluoro-2-propanol, derivatization grade (GC derivatization), LiChropur, ≥99.8%