MilliporeSigma
  • Calcium antagonist added to angiotensin receptor blocker: a recipe for reducing blood pressure variability?: evidence from day-by-day home blood pressure monitoring.
Calcium antagonist added to angiotensin receptor blocker: a recipe for reducing blood pressure variability?: evidence from day-by-day home blood pressure monitoring.
Hypertension (Dallas, Tex. : 1979) (2012-05-02)
Gianfranco Parati, Grzegorz Bilo
Sigma-Aldrich
Azelnidipine, ≥98% (HPLC), powder