MilliporeSigma
  • A novel one-pot pseudo-five-component condensation reaction towards bifunctional diazepine-tetrazole containing compounds: synthesis of 1H-tetrazolyl-1H-1,4-diazepine-2,3-dicarbonitriles and 1H-tetrazolyl-benzo[b][1,4]diazepines.
A novel one-pot pseudo-five-component condensation reaction towards bifunctional diazepine-tetrazole containing compounds: synthesis of 1H-tetrazolyl-1H-1,4-diazepine-2,3-dicarbonitriles and 1H-tetrazolyl-benzo[b][1,4]diazepines.
Molecular diversity (2012-05-18)
Hamid Mofakham, Ahmad Shaabani, Sajjad Mousavifaraz, Fatemeh Hajishaabanha, Shabnam Shaabani, Seik Weng Ng
ABSTRACT
A novel and efficient method has been developed for the one-pot synthesis of bifunctional diazepine-tetrazole containing compounds. 1H-Tetrazolyl-1H-1,4-diazepine-2, 3-dicarbonitrile and 1H-tetrazolyl-benzo[b][1,4]diazepine derivatives were synthesized in good yields using 2,3-diaminomaleonitrile or an aromatic diamine, ketones, trimethylsilyl azide, and an isocyanide in the presence of p-toluenesulfonic acid as a catalyst in methanol at room temperature.
Sigma-Aldrich
Tetrazole solution, suitable for DNA synthesis, filtered through a 1 μm filter, ~0.45 M in acetonitrile