MilliporeSigma
  • Famotidine suppresses osteogenic differentiation of tendon cells in vitro and pathological calcification of tendon in vivo.
Famotidine suppresses osteogenic differentiation of tendon cells in vitro and pathological calcification of tendon in vivo.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2012-05-18)
Kenichi Yamamoto, Hironori Hojo, Isao Koshima, Ung-il Chung, Shinsuke Ohba
ABSTRACT
Heterotopic ossification or calcification follows any type of musculoskeletal trauma and is known to occur after arthroplasties of hip, knee, shoulder, or elbow; fractures; joint dislocations; or tendon ruptures. Histamine receptor H2 (Hrh2) has been shown to be effective for reducing pain and decreasing calcification in patients with calcifying tendinitis, which suggested that H2 blockers were effective for the treatment of tendon ossification or calcification. However, the detailed mechanisms of its action on tendon remain to be clarified. We investigated the mechanisms underlying H2 blocker-mediated suppression of tendon calcification, with a focus on the direct action of the drug on tendon cells. Famotidine treatment suppressed the mRNA expressions of Col10a1 and osteocalcin, ossification markers, in a tendon-derived cell line TT-D6, as well as a preosteoblastic one MC3T3-E1. Both of the cell lines expressed Hrh2; histamine treatment induced osteocalcin expression in these cells. Famotidine administration suppressed calcification in the Achilles tendon of ttw mice, a mouse model of ectopic ossification. These data suggest that famotidine inhibits osteogenic differentiation of tendon cells in vitro, and this inhibition may underlie the anti-calcification effects of the drug in vivo. This study points to the use of H2 blockers as a promising strategy for treating heterotopic ossification or calcification in tendon, and provides evidence in support of the clinical use of famotidine.
Product Description
Supelco
Famotidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Famotidine