MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Self-catalytic degradation of ortho-chlorophenol with Fenton's reagent studied by chemiluminescence.
Self-catalytic degradation of ortho-chlorophenol with Fenton's reagent studied by chemiluminescence.
Journal of environmental sciences (China) (2012-06-05)
Zhen Lin, Hui Chen, Yun Zhou, Nobuaki Ogawa, Jin-Ming Lin
ABSTRACT
The degradation of ortho-chlorophenol using Fenton's reagent was studied by chemiluminescence (CL). Without a special CL reagent, a weak CL emission from the mixture of ferrous ion and hydrogen peroxide was observed at room temperature. The CL intensity was increased by the addition of ortho-chlorophenol into the mixed solution. When the temperature was raised to 65 degrees C, the CL intensity was enhanced strongly. The CL mechanisms for the system H2O2-Fe2+ with and without ortho-chlorophenol were studied by examining the CL spectrum, gas chromatography-mass spectrometry and electron spin resonance spectrum. The effects of various free radical scavengers, surfactants and fluorescence compounds on the CL intensity were also investigated. A self-catalytic oxidation mechanism was proposed. The results showed that singlet oxygen was the main emitter for the system H2O2-Fe2+. The strong CL from the system H2O2-Fe(2+)-ortho-chlorophenol was due to singlet oxygen and electronically excited quinone. The benzenediol-like intermediate product formed during the phenol oxidation process greatly promoted the Fenton's reaction and led to higher CL intensity. Chemiluninescence is a novel approach for the investigation of the oxidation of some organic pollutants by Fenton's reagent.
Sigma-Aldrich
2-Chlorophenol, SAJ first grade, ≥99.0%
Sigma-Aldrich
2-Chlorophenol, ≥99%
Sigma-Aldrich
2-Chlorophenol, 98%