MilliporeSigma
  • Approach to the synthesis of indoline derivatives from diaryliodonium salts.
Approach to the synthesis of indoline derivatives from diaryliodonium salts.
The Journal of organic chemistry (2012-06-08)
Kamalkishor P Landge, Keun Sam Jang, Sang Yeul Lee, Dae Yoon Chi
ABSTRACT
An effective method of constructing the indoline moiety via intramolecular nucleophilic ring closure of a diaryliodonium salt is described. Diacetoxyiodoarene compounds (1a-1e) were converted into intermediate Koser's reagent and coupled with arylstannanes (7-10) to form diaryliodonium salts (11a-14e). Indoline compounds with different N-protecting groups, 15, 16, 17, and 18, were synthesized in higher yields by treating salts (11a-14e) with Cs(2)CO(3) and TEMPO. Regardless of the electronic environment of five para-substituted iodoarenes and the natures of four N-protected arylstannane groups, the conversion proceeded well to afford corresponding indolines in yields of 72-84 and 70-84%, respectively.
Sigma-Aldrich
Indoline, ReagentPlus®, 99%