MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
A silver-catalyzed spirocyclization of alkynyl silyl enol ethers.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-06-08)
Christian Schäfer, Michel Miesch, Laurence Miesch
ABSTRACT
Ring out the old: The cycloisomerization of alkynyl silyl enol ethers proceeds at ambient temperature under the mild conditions of silver catalysis. Mono- or bicyclic spiro compounds can be obtained by 5-exo-dig reactions. Trapping the vinyl silver species with an iodide source, such as N-iodosuccinimide (NIS), afforded the alkenyl iodide derivatives.
Sigma-Aldrich
N-Iodosuccinimide, 95%