MilliporeSigma
  • Ultraviolet-ultraviolet hole burning spectroscopy in a quadrupole ion trap: dibenzo[18]crown-6 complexes with alkali metal cations.
Ultraviolet-ultraviolet hole burning spectroscopy in a quadrupole ion trap: dibenzo[18]crown-6 complexes with alkali metal cations.
Angewandte Chemie (International ed. in English) (2012-06-13)
Chang Min Choi, Dae Ho Choi, Jiyoung Heo, Nam Joon Kim, Seong Keun Kim
ABSTRACT
Conformation selective: A new technique of ultraviolet-ultraviolet hole burning spectroscopy that can be applied to ions stored in a quadrupole ion trap (QIT) is developed and used to obtain the conformation-selective electronic spectra of dibenzo[18]crown-6 complexes with alkali metal cations (M(+), see picture; F(+) = fragment).
Sigma-Aldrich
Dibenzo-18-crown-6, 98%