MilliporeSigma
  • Technical note: Hapten synthesis, antibody production and development of an enzyme-linked immunosorbent assay for detection of the natural steroidal alkaloid Dendrogenin A.
Technical note: Hapten synthesis, antibody production and development of an enzyme-linked immunosorbent assay for detection of the natural steroidal alkaloid Dendrogenin A.
Biochimie (2012-06-20)
Philippe de Medina, Michael R Paillasse, Gregory Segala, Talal Al Saati, Jeannine Boyes, Georges Delsol, Cuider Allal, Sabrina Marsili, Sandrine Silvente-Poirot, Marc Poirot
ABSTRACT
We have recently discovered the existence of 5α-Hydroxy-6β-[2-(1H-imidazol-4-yl)ethylamino]cholestan-3β-ol, called Dendrogenin A (DDA), as the first endogenous steroidal alkaloid ever described in mammals. We found that the DDA content of tumors and cancer cell lines was low or absent compared with normal cells showing that a deregulation in DDA biosynthesis was associated with cancer and therefore suggesting that DDA could represent a metabolomic cancer biomarker. This prompted us to produce antibodies that selectively recognize DDA. For this purpose, the hapten 5α-hydroxy-6β-[2-(1H-imidazol-4-yl)ethylamino]cholestan-3β-o-hemisuccinate with a carboxylic spacer arm attached to the 3β-hydroxyl group of DDA was synthesized. The hapten was coupled to bovine serum albumin and keyhole limpet hemocyanin for antibody production to develop an enzyme-linked immunosorbent assay (ELISA). The protein conjugates were injected into BALB/c mice to raise antibodies. The monoclonal antibodies that were secreted from the hybridoma cell lines established were assessed with indirect ELISA by competitive assays using dilutions of a DDA standard. The antibodies from the selected hybridomas had an IC(50) value ranging from 0.8 to 425 ng/ml. Three antibodies showed no cross-reactivity with structurally related compounds including histamine, cholesterol, ring B oxysterols and a regio-isomer of DDA. In this study, high-affinity and selective antibodies against DDA were produced for the first time, and a competitive indirect ELISA was developed.
Sigma-Aldrich
5β-Cholestan-3α-ol, ≥95%
Sigma-Aldrich
5α-Cholestan-3β-ol, ≥95%
Sigma-Aldrich
Coprostan-3-ol, ≥98%