MilliporeSigma
  • [1-Adamanty fluoroformate, a useful reagent in peptide chemistry (author's transl)].
[1-Adamanty fluoroformate, a useful reagent in peptide chemistry (author's transl)].
Hoppe-Seyler's Zeitschrift fur physiologische Chemie (1976-11-01)
L Moroder, L Wackerle, E Wünsch
ABSTRACT
1-Adamantyl fluoroformate was prepared from 1-adamantol and fluorophosgene. The cristalline and stable fluoroformate was allowed to react with amino acids to give the corresponding 1-adamantyloxycarbonyl derivatives in consistently high yields. Additonally this new reagent proved to be ideally reactive for acylation of the imidazol function of histidine and histidyl-peptides.