MilliporeSigma
  • Preparation and characterization of porous cross linked laccase aggregates for the decolorization of triphenyl methane and reactive dyes.
Preparation and characterization of porous cross linked laccase aggregates for the decolorization of triphenyl methane and reactive dyes.
Bioresource technology (2012-06-26)
V Vinoth Kumar, M P Prem Kumar, K V Thiruvenkadaravi, P Baskaralingam, P Senthil Kumar, S Sivanesan
ABSTRACT
The production of porous cross-linked enzyme aggregates (p-CLEAs) is a simple and effective methodology for laccase immobilization. A three-phase partitioning technique was applied to co-precipitate laccase and starch, followed by cross-linking with glutaraldehyde and removal of starch by α-amylase to create pores in the CLEAs. Scanning electron microscopy revealed a very smooth spherical structure with numerous large pores. The half-life of free laccase at 55°C was calculated to be 1.3h, while p-CLEAs did not lose any activity even after 14 h. p-CLEAs also exhibited improved storage stability, catalytic efficiency and could be recycled 15 times with 60% loss of activity. The catalysts decolorized triphenylmethane and reactive dyes by 60-70% at initial dye concentrations of 2 and 0.5 g L(-1), respectively, without any mediators. These results suggest the potential of CLEA technology in waste-water treatment.
Sigma-Aldrich
Triphenylmethane, 99%