MilliporeSigma
  • Transition metal-catalyzed fluorination of multi carbon-carbon bonds: new strategies for fluorinated heterocycles.
Transition metal-catalyzed fluorination of multi carbon-carbon bonds: new strategies for fluorinated heterocycles.
Organic & biomolecular chemistry (2012-06-27)
Guosheng Liu
ABSTRACT
Transition metal catalysts have been developed for introducing fluorine into organic compounds. Recent progress in this area is reviewed with an emphasis on the selective fluorination of alkenes, alkynes and allenes. Regio- and stereoselective fluorination reactions are highlighted.
Sigma-Aldrich
Allene, ≥95%