MilliporeSigma
Long-term toxicology and carcinogenicity of 2,4,6-trichlorophenol.
Chemosphere (2012-07-04)
James Huff
ABSTRACT
Carcinogenesis bioassays were conducted by giving 2,4,6-trichlorophenol [2,4,6-TCP] in feed to groups of 50 male and female Fischer rats and male B6C3F1 mice for two years. Dietary concentrations were 0 [20/group], 5000 [0.5%], or 10,000 [1%] ppm. Female mice began with 10,000 and 20,000 ppm but after 38 weeks were lowered due to reduced body weights to 2500 and 5000 ppm for 67 weeks; exposures averaged 5200 and 10,400 ppm. Adverse effects at two years were leukocytosis and monocytosis of peripheral blood and hyperplasia of bone marrow in both sexes of rats. In mice, liver toxicity, including individual liver cell abnormalities, focal areas of cellular alteration, and focal and nodular areas of hyperplasia were commonly present. Regarding carcinogenic activity, TCP caused leukemias/lymphomas in male rats, and possibly in female rats and female mice as well, and induced liver tumors in male and female mice. Using NTP categories of evidence indicates 'clear evidence of carcinogenicity' for male rats [hematopoietic system tumors]; 'equivocal evidence of carcinogenicity' for female rats [hematopoietic system tumors]; 'clear evidence of carcinogenicity' for male and female mice [liver tumors].
Sigma-Aldrich
2,4,6-Trichlorophenol, 98%