MilliporeSigma
  • The protective role of oxymatrine on neuronal cell apoptosis in the hemorrhagic rat brain.
The protective role of oxymatrine on neuronal cell apoptosis in the hemorrhagic rat brain.
Journal of ethnopharmacology (2012-07-04)
Man Huang, Yue-Yu Hu, Xiao-Qiao Dong, Qiu-Ping Xu, Wen-Hua Yu, Zu-Yong Zhang
ABSTRACT
Oxymatrine is extracted from the traditional Chinese herb Sophora flavescens Ait, possesses anti-inflammatory, anti-oxidative and anti-apoptotic properties, and has been used for the treatment of chronic viral hepatitis and many other diseases. This study aimed to investigate the effects of oxymatrine on inflammatory response mediated by Toll-like receptor4 (TLR4) and nuclear factor kappa-B (NF-κB), oxidative injury induced by 12/15 lipoxygenase (12/15-LOX), phosphorylated p38 mitogen activated protein kinase (phosphor-p38 MAPK) and cytosolic phospholipase A2 (cPLA2), and neuronal cell apoptosis in rat brain with intracerebral hemorrhage (ICH). Wistar rats were treated intraperitoneally with 60 or 120mg/kg of oxymatrine daily for 5 days following ICH. The rats were sacrificed at hour 2, 6, 12, 24, 48, 72, and 120 after ICH. The gene expressions of TLR-4 and NF-κB, the levels of TNF-alpha, interleukin-1beta, interleukin-6, 12/15-LOX, phospho-p38 MAPK and cPLA2, and the number of apoptotic neuronal cells in rat brain were determined. Oxymatrine at 120mg/kg significantly suppressed gene expressions of TLR-4 and NF-κB, decreased levels of TNF-alpha, interleukin-1beta and interleukin-6, inhibited synthesis of 12/15-LOX, phospho-p38 MAPK and cPLA2 protein, and mitigated apoptotic neuronal changes following ICH in rat. Oxymatrine at 120mg/kg following ICH inhibits inflammatory responses, oxidative injury, and neuronal cell apoptosis in rats.