MilliporeSigma
  • Comparative evaluation of selenium accumulation by allium species after foliar application of selenium nanoparticles, sodium selenite and sodium selenate.
Comparative evaluation of selenium accumulation by allium species after foliar application of selenium nanoparticles, sodium selenite and sodium selenate.
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections (2012-07-05)
N A Golubkina, G E Folmanis, I G Tananaev
ABSTRACT
Accumulation and translocation intensity of selenium nanoparticles by natural selenium accumulators-perennial onions-is established and a decrease of these parameters is demonstrated in a sequence: Se(+6) > Se(0) > Se(+4).
Sigma-Aldrich
Sodium selenate, ≥95% (elemental analysis)
Sigma-Aldrich
Sodium selenate, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Sodium selenate, BioXtra