MilliporeSigma
  • Surface enhancement of WO3 nanowires toward the oxidation and electrochemical detection of honokiol in traditional Chinese medicine.
Surface enhancement of WO3 nanowires toward the oxidation and electrochemical detection of honokiol in traditional Chinese medicine.
Colloids and surfaces. B, Biointerfaces (2012-07-07)
Wanyun Qu, Xiaoyong Xiong, Weibing Hu, Pan Zhang, Qi Luo, Shenghui Zhang
ABSTRACT
WO(3) nanowires (nano-WO(3)) were synthesized using a simple hydrothermal technique, and then used to modify the carbon paste electrode (CPE). The electrochemical behavior of honokiol on the unmodified CPE and nano-WO(3) modified CPE was compared. It was found that nano-WO(3) remarkably increased the oxidation signal of honokiol. The influences of supporting electrolyte, amount of nano-WO(3), accumulation potential and time were studied on the oxidation peak current of honokiol. Based on the strong enhancement effect of nano-WO(3), a sensitive, simple and rapid electrochemical method was developed for the detection of honokiol. The linear range was from 3×10(-8) to 2.0×10(-5)M, and the detection limit was as low as 1×10(-8)M after 2-min accumulation. Finally, it was used to determine honokiol in traditional Chinese medicines, and the recovery was over the range between 96.5% and 98.8%.
Sigma-Aldrich
Honokiol, ≥98% (HPLC), powder