MilliporeSigma
  • Biological activities of the polysaccharides produced in submerged culture of two edible Pleurotus ostreatus mushrooms.
Biological activities of the polysaccharides produced in submerged culture of two edible Pleurotus ostreatus mushrooms.
Journal of biomedicine & biotechnology (2012-07-11)
Emanuel Vamanu
ABSTRACT
Exopolysaccharides (EPS) and internal (intracellular) polysaccharides (IPS) obtained from the Pleurotus ostreatus M2191 and PBS281009 cultivated using the batch system revealed an average of between 0.1-2 (EPS) and 0.07-1.5 g/L/day (IPS). The carbohydrate analysis revealed that the polysaccharides comprised 87-89% EPS and 68-74% IPS. The investigation of antioxidant activity in vitro revealed a good antioxidant potential, particularly for the IPS and EPS isolated from PBS281009, as proved by the EC(50) value for DPPH, ABTS scavenging activity, reducing power, and iron chelating activity.
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p′-disulfonic acid monosodium salt hydrate, 97%
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, Indicator ligand
Supelco
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, for spectrophotometric det. of Fe, 97.0%
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, BioXtra