MilliporeSigma
  • The microstructure of collagen type I gel cross-linked with gold nanoparticles.
The microstructure of collagen type I gel cross-linked with gold nanoparticles.
Colloids and surfaces. B, Biointerfaces (2012-07-17)
Thomas Schuetz, Nathan Richmond, Marianne E Harmon, Joseph Schuetz, Luciano Castaneda, Katarzyna Slowinska
ABSTRACT
Scanning electron microscopy, transmission electron microscopy, rheometry, and electrochemistry were used to provide insight into the microstructure of collagen type I gel (1%, w/v) modified with the tiopronin-protected (N-(2-mercaptopropionyl)glycine) gold nanoparticles (TPAu), a multivalent crosslinker. The cross-linking reaction, performed via EDC (1-ethyl-3-(3-dimethyl aminopropyl) carbodiimide) coupling, results in compliant, mechanically stable and continuous gels. The gels contain unusual interconnected collagen-TPAu particles. Electrochemical measurements of 4-hydroxy-(2,2,6,6-tetramethylpiperidine-1-oxyl) (4HT) diffusion within the gel reveal that the gel hindrance is nearly independent of the TPAu concentration. The properties of the collagen-TPAu gel make it suitable for potential biomedical applications, such as delivery of small molecule drugs.
Sigma-Aldrich
N-(2-Mercaptopropionyl)glycine