MilliporeSigma
  • On-line (HPLC-NMR) and off-line phytochemical profiling of the Australian plant, Lasiopetalum macrophyllum.
On-line (HPLC-NMR) and off-line phytochemical profiling of the Australian plant, Lasiopetalum macrophyllum.
Natural product communications (2012-07-18)
Michael Timmers, Sylvia Urban
ABSTRACT
On-line (HPLC-NMR) and off-line (HPLC, NMR and MS) methodologies were used to profile the constituents present in the crude extract of Lasiopetalum macrophyllum. On-flow and stop-flow HPLC-NMR supported the presence of trans-tiliroside and permitted partial identification of cis-tiliroside and 4'-methoxy-trans-tiliroside. Off-line isolation led to the unequivocal identification of four flavanoid glycosides including a new structural derivative, 4'-methoxycis-tiliroside. This is the first report of flavonoid glycosides occurring in this plant genus. In addition, a number of structure revisions have been proposed for previously reported flavonoid glycosides that were incorrectly assigned.