MilliporeSigma
  • Protonated carbonic acid and reactive intermediates in the acidic decarboxylation of indolecarboxylic acids.
Protonated carbonic acid and reactive intermediates in the acidic decarboxylation of indolecarboxylic acids.
The Journal of organic chemistry (2012-07-19)
Adelle A Vandersteen, Scott O C Mundle, Ronald Kluger
ABSTRACT
Elucidation of the mechanism for decarboxylation of indolecarboxylic acids over a wide range of solution acidity reveals the importance of protonated carbonic acid (PCA) as a reaction intermediate. In concentrated acid, the initial addition of water to the carboxyl group of the indolecarboxylic acid leads to a hydrated species that is capable of releasing PCA upon rate-determining carbon-carbon bond cleavage. The overall process is catalytic in water and acid, implicating PCA as a potential carboxylating reagent in the microscopic reverse reaction.
Sigma-Aldrich
Indole-2-carboxylic acid, 98%