MilliporeSigma
  • A single-step kit formulation for the (99m)Tc-labeling of HYNIC-Duramycin.
A single-step kit formulation for the (99m)Tc-labeling of HYNIC-Duramycin.
Nuclear medicine and biology (2012-08-04)
Ming Zhao, Zhixin Li
ABSTRACT
(99m)Tc-Duramycin is a unique radiopharmaceutical that binds specifically to phosphatidylethanolamine (PE). The current effort is to develop a single-step kit formulation for the (99m)Tc labeling of HYNIC-Duramycin. A titration series of Tricine/TPPTS coligand systems were tested for an optimal formulation to produce (99m)Tc-Duramycin with high radiochemical purity and specific activity. The radiopharmaceutical prepared using the kit formulation was tested for PE binding specificity using polystyrene microbeads coated with different phospholipid species. Radiochemical performance of the kits was assessed after storage at -20°C, room temperature and 37°C. Biodistribution profile of kit-prepared (99m)Tc-Duramycin was characterized in healthy rats at 3, 10, 20, 60 and 180min after intravenous injection. Binding studies were performed using the rat aortic arch and a rat model of myocardial ischemia/reperfusion, which represent scenarios of physiological and pathological PE externalization. A Tricine/TPPTS ratio of 10:1 led to a consistent production of (99m)Tc-Duramycin with high radiochemical purity (> 90%), whereas a higher ratio at 40:1 produced radiopharmaceuticals with incomplete substitution of Tricine coligand. (99m)Tc-Duramycin prepared using the single-step kit formulation retained PE-binding specificity. The kits are stable over long-term storage. The biodistribution profile of kit-prepared (99m)Tc-Duramycin is consistent with HPLC purified radiopharmaceutical from prior studies. Binding studies on a tissue level indicate that the radiopharmaceutical is suitable for studying biological processes that involve PE distribution and redistribution in various physiological and pathological conditions. A single-step kit formulation is developed for (99m)Tc-labeling of HYNIC-Duramycin. The radiopharmaceutical has high radiochemical purity and specific activity, retained PE binding activities, amiable to long-term storage, and is injection-ready for in vivo applications.
Sigma-Aldrich
Duramycin from Streptoverticillium cinnamoneus, ≥90.0%