MilliporeSigma
Does Microcystis aeruginosa have quorum sensing?
FEMS microbiology letters (2012-08-07)
Chunmei Zhai, Ping Zhang, Fei Shen, Changxin Zhou, Changhong Liu
ABSTRACT
Quorum sensing (QS) is a system of cell-to-cell communication by means of intercellular signaling molecules to coordinate a set of targeted gene expression or repression in many Gram-negative bacteria; it plays important roles for bacteria in adaptation to adverse environmental conditions. In this study, we first demonstrated that Microcystis aeruginosa PCC-7820 could produce QS-related signal acylated homoserine lactones (AHLs) among the metabolite of axenic M. aeruginosa, based on bioassay and liquid chromatography-mass spectrometry (LC-MS) analysis. The concentration of the AHLs in the culture medium was cell density dependent and reached a maximum of 18 nM at 1.03 × 10(7) cells mL(-1), 30 days after inoculation. The regulation mechanism of QS in M. aeruginosa and its possible role in bloom formation are discussed.
Sigma-Aldrich
(S)-α-Amino-γ-butyrolactone hydrochloride, 97%
Sigma-Aldrich
α-Amino-γ-butyrolactone hydrobromide, 99%
Sigma-Aldrich
L-Homoserine lactone hydrochloride
Sigma-Aldrich
Dihydro-3-amino-2-(3H)-furanone, AldrichCPR