MilliporeSigma
  • Colestyramine slows gastric emptying of liquids and reduces appetite in healthy subjects.
Colestyramine slows gastric emptying of liquids and reduces appetite in healthy subjects.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society (2012-08-07)
A Psichas, T Little, S Lal, J McLaughlin
ABSTRACT
There is evidence to suggest that the particulate resin colestyramine, a bile acid sequestrant formerly used as a cholesterol-lowering agent, enhances secretion of the gut hormone cholecystokinin (CCK). Established physiological actions of CCK include inhibition of gastric emptying and induction of satiation. This study evaluated the hypothesis that colestyramine, which is luminally retained, would slow gastric emptying of liquids and suppress appetite in humans. Nine healthy volunteers consumed 500 mL liquid test meals containing 4 g colestyramine, 12 g colestyramine, or control (water alone), on three occasions, in a randomized order. The effect of colestyramine on gastric emptying was determined non-invasively using the (13) C-acetate breath test, and appetite and other gut-centered sensations were rated using visual analog scale questionnaires. Colestyramine dose dependently slowed liquid gastric emptying compared with control (water) (4 g vs control, ∼20% reduction, P < 0.05; 12 g vs control, ∼35% reduction, P < 0.01). Colestyramine also significantly reduced hunger (4 g vs control, ∼20% reduction, P < 0.01), and the amount of food participants felt able to eat (12 g vs control, ∼32% reduction, P < 0.001), but increased bloating (both doses, P < 0.05), with no effect on ratings of nausea. This study provides the first evidence that colestyramine significantly slows liquid gastric emptying and reduces appetite in healthy humans. Colestyramine therefore presents an attractive gut-brain signaling research tool in that it is not absorbed and thus lacks potentially confounding postabsorptive effects. Furthermore, with clear effects on gastric emptying and appetite, colestyramine now merits consideration as a trial therapeutic strategy for appetite suppression and weight loss.
Sigma-Aldrich
Cholestyramine resin