MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Pharmacokinetics and toxicity of antimuscarinic drugs for overactive bladder treatment in females.
Pharmacokinetics and toxicity of antimuscarinic drugs for overactive bladder treatment in females.
Expert opinion on drug metabolism & toxicology (2012-08-09)
Umberto Leone Roberti Maggiore, Stefano Salvatore, Franco Alessandri, Valentino Remorgida, Massimo Origoni, Massimo Candiani, Pier Luigi Venturini, Simone Ferrero
ABSTRACT
Antimuscarinics (AMs) are the mainstay of pharmacological treatment of overactive bladder (OAB), a symptom complex defined by the presence of urinary urgency, usually associated with frequency and nocturia, with or without urgency urinary incontinence. The AMs used to treat OAB differ in their pharmacological profiles, which may affect their potential for causing adverse effects (AEs). The present article aims to review the literature about pharmacokinetics (PK) of the different AMs used in the treatment of OAB. Furthermore, the AEs related to the use of these drugs and their incidence are presented. This systematic review is based on material searched and obtained via Medline, Pubmed and EMBASE up to March 2012 using the search terms "adverse events, pharmacokinetics, tolerability" in combination with "darifenacin, fesoterodine, imidafenacin, oxybutynin, propiverine, solifenacin, tolterodine, and trospium." Antimuscarinics are the first-line pharmacological treatment for OAB. Despite the development of new molecules that improve their efficacy/safety profile, there are some drugs that are pharmacokinetically more appropriate to be prescribed in specific populations such as patients with neurological disease or the elderly. Moreover, research should be encouraged in evaluating antimuscarinics in conjunction with other drugs such as estrogens or beta-agonists. The identification of prognostic criteria for pharmacological therapy would be helpful.
Sigma-Aldrich
Oxybutynin chloride, meets EP, USP testing specifications
Sigma-Aldrich
Oxybutynin chloride, ≥98% (TLC), powder
Sigma-Aldrich
Propiverine hydrochloride, ≥98% (HPLC)