MilliporeSigma
  • Residues of organochlorine pesticides in water and suspended particulate matter from Xiangshan Bay, East China Sea.
Residues of organochlorine pesticides in water and suspended particulate matter from Xiangshan Bay, East China Sea.
Bulletin of environmental contamination and toxicology (2012-08-10)
Weidong Li, Huayun Yang, Qinfen Gao, Huanbo Pan, Haobo Yang
ABSTRACT
Residual levels, distribution and possible sources of organochlorine pesticides (OCPs) in nine water column and suspended particulate matter (SPM) samples collected from Xiangshan Bay, East China Sea were investigated. Total OCPs concentrations in water column and SPM ranges from 2.88 to 34.72 ng/L and 2.47 to 29.94 ng/L, respectively, which exceed the US Environmental Protection Agency regulatory limits. Moreover, low ratio of α-HCH/γ-HCH and high ratios β- and γ-HCH suggested the cocktail input pattern of fresh and weathered HCHs. Meanwhile, low ratio of (DDE + DDD)/DDT and high ratios p,p'- and o,p'-DDT reflected a "dicofol type DDT pollution" pattern.