MilliporeSigma
  • Identification of new ligands for the methionine biosynthesis transcriptional regulator (MetJ) by FAC-MS.
Identification of new ligands for the methionine biosynthesis transcriptional regulator (MetJ) by FAC-MS.
Journal of molecular microbiology and biotechnology (2012-08-15)
Ricardo Martí-Arbona, Munehiro Teshima, Penelope S Anderson, Kristy L Nowak-Lovato, Elizabeth Hong-Geller, Clifford J Unkefer, Pat J Unkefer
ABSTRACT
We have developed a high-throughput approach using frontal affinity chromatography coupled to mass spectrometry (FAC-MS) for the identification and characterization of the small molecules that modulate transcriptional regulator (TR) binding to TR targets. We tested this approach using the methionine biosynthesis regulator (MetJ). We used effector mixtures containing S-adenosyl-L-methionine (SAM) and S-adenosyl derivatives as potential ligands for MetJ binding. The differences in the elution time of different compounds allowed us to rank the binding affinity of each compound. Consistent with previous results, FAC-MS showed that SAM binds to MetJ with the highest affinity. In addition, adenine and 5'-deoxy-5'-(methylthio)adenosine bind to the effector binding site on MetJ. Our experiments with MetJ demonstrate that FAC-MS is capable of screening complex mixtures of molecules and identifying high-affinity binders to TRs. In addition, FAC-MS experiments can be used to discriminate between specific and nonspecific binding of the effectors as well as to estimate the dissociation constant (K(d)) for effector-TR binding.
Sigma-Aldrich
5′-Deoxy-5′-(methylthio)adenosine